missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C19orf44 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583966

Product Code. 16390915

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SEVSEHLSAS SASAIQQDST SSMQPPSEAP MVNTVSSAYS EDFENSPSLT ASEPTAHSKE SLDRTLDALS ESSSSVKTDL PQTAESRK Highest antigen sequence indentity to the following orthologs: Rat - 42%, Mouse - 41%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C19orf44
Polyclonal
Unconjugated
C19orf44
C19orf44; chromosome 19 open reading frame 44; FLJ21742; RGD1311847; similar to 1700030K09Rik protein; uncharacterized protein C19orf44; Uncharacterized protein C19orf44 homolog
Rabbit
Antigen affinity chromatography
RUO
84167
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9H6X5
C19orf44
Recombinant protein corresponding to Human C19orf44. Recombinant protein control fragment (Product #RP-104870).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.