missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19ORF80 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808682S
This item is not returnable.
View return policy
Description
C19ORF80 Monoclonal antibody specifically detects C19ORF80 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| C19ORF80 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UM19 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51440 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B12 | |
| Unconjugated | |
| HPCAL4 | |
| angiopoietin like 8; angiopoietin-like 8; angiopoietin-like protein 8; Angptl8; betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80; EG624219; Gm6484; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; hepatocellular carcinoma-associated protein TD26 homolog; lipasin; LOC100361444; predicted gene 6484; PRO1185; PVPA599; rCG31799-like; refeeding-induced fat and liver protein; Rifl; TD26; UNQ599/PRO1185 | |
| Human recombinant protein fragment corresponding to amino acids 2-191 of human HPCAL4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction