missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C1GALT1C1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15741291 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15741291 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15741291 Supplier Invitrogen™ Supplier No. PA553349

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NRMHHHEHHH LQAPNKEDIL KISEDERMEL SKSFRVYCII LVKPKDVSLW AAVKETWTKH CDKAEFFSSE NVKVFESINM DTNDMWLMMR KAYKYAFDKY RDQYNWFFLA RPTTFAIIE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1GALT1C1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1GALT1C1
Gene Accession No. Q96EU7
Gene Alias 1500002I11Rik; AB030184; beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1GALT1 specific chaperone 1; C1galt1c1; C1GALT1-specific chaperone 1; C1GALT2; C1Gal-T2; C38H2-L1; C38H2-like protein 1; C81205; core 1 beta1,3-galactosyltransferase 2; core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 beta3-Gal-T2; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; COSMC; HSPC067; mC1Gal-T2; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310
Gene Symbols C1GALT1C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1GALT1C1. Recombinant protein control fragment (Product #RP-92239).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29071
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.