missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf102 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555849
This item is not returnable.
View return policy
Description
Immunogen sequence: TMSVRTLPLL FLNLGGEMLY ILDQRLRAQN IPGDKARKVL NDIISTMFNR KFMEELFKPQ ELYSKKALRT VYERLAHASI MKLNQASMDK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
May be involved in drug clearance in the placenta. [UniProt].
Specifications
| C1orf102 | |
| Polyclonal | |
| Unconjugated | |
| OSCP1 | |
| 1810007P19Rik; 5730415O04; 6030436A01Rik; C1orf102; hOSCP1; NOR1; organic solute carrier 1; organic solute carrier partner 1; organic solute carrier protein 1; organic solute transport protein 1; Oscp1; oxidored nitro domain containing protein; oxidored-nitro domain-containing protein; oxidored-nitro domain-containing protein 1; Protein OSCP1; RGD1306596; similar to oxidored-nitro domain-containing protein isoform 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127700 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WVF1 | |
| OSCP1 | |
| Recombinant protein corresponding to Human C1orf102. Recombinant protein control fragment (Product #RP-94096). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction