missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf104 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570026
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 91%; Human: 100%.
RUSC1-AS1 is a protein coding gene.
Specifications
| C1orf104 | |
| Polyclonal | |
| Unconjugated | |
| RUSC1-AS1 | |
| C1orf104; putative uncharacterized protein RUSC1-AS1; RUSC1 antisense gene protein 1; RUSC1 antisense RNA 1; RUSC1 antisense RNA 1 (non-protein coding); RUSC1-AS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 284618 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q66K80 | |
| RUSC1-AS1 | |
| synthetic peptide directed towards the middle region of human C1orf104. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction