missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf106 Monoclonal Antibody (OTI14A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806347S
This item is not returnable.
View return policy
Description
C1orf106 Monoclonal antibody specifically detects C1orf106 in Human samples. It is validated for Western BlotSpecifications
| C1orf106 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15466 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8431 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI14A10 | |
| Unconjugated | |
| NR0B2 | |
| 1700034M08Rik; 4933426C09Rik; 5730559C18Rik; AI586180; C1orf106; chromosome 1 open reading frame 106; D1Mgi54; Inava; Innate immunity activator protein; RGD1311892; RIKEN cDNA 5730559C18 gene; similar to hypothetical protein FLJ10901; uncharacterized protein C1orf106; uncharacterized protein C1orf106 homolog | |
| Full length human recombinant protein of human NR0B2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction