missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf111 Monoclonal Antibody (OTI1A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811093S
This item is not returnable.
View return policy
Description
C1orf111 Monoclonal antibody specifically detects C1orf111 in Human samples. It is validated for Western BlotSpecifications
| C1orf111 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5T0L3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 284680 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1A9 | |
| Unconjugated | |
| SPATA46 | |
| 1700015E13Rik; C1orf111; chromosome 1 open reading frame 111; HSD20; RGD1565283; RIKEN cDNA 1700015E13 gene; RP11-565P22.3; similar to novel protein; SPATA46; Spermatogenesis-associated protein 46; uncharacterized protein C1orf111; uncharacterized protein C1orf111 homolog | |
| Full length human recombinant protein of human C1ORF111 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction