Learn More
C1orf111 Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811094S
Encompass_Preferred
 View more versions of this product
Product Code. 17504744
  • € 242.00 / 30µL
Description
C1orf111 Monoclonal antibody specifically detects C1orf111 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
C1orf111
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5T0L3
Mouse
Affinity chromatography
RUO
284680
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2F3
Unconjugated
SPATA46
1700015E13Rik; C1orf111; chromosome 1 open reading frame 111; HSD20; RGD1565283; RIKEN cDNA 1700015E13 gene; RP11-565P22.3; similar to novel protein; SPATA46; Spermatogenesis-associated protein 46; uncharacterized protein C1orf111; uncharacterized protein C1orf111 homolog
Full length human recombinant protein of human C1ORF111 produced in E.coli.
30 μL