Learn More
C1orf111 Monoclonal Antibody (OTI2F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811094
Encompass_Preferred
 View more versions of this product
Product Code. 17584734
  • € 540.87 / 100µL
Description
Play a role in spermiogenesis and fertilization.
Specifications
Specifications
C1orf111
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5T0L3
SPATA46
Full length human recombit protein of human C1ORF111 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2F3
Unconjugated
SPATA46
C1orf111, HSD20
Mouse
Affinity Chromatography
RUO
284680
-20° C, Avoid Freeze/Thaw Cycles