Learn More
Invitrogen™ C1orf123 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113398
Product Code. 17191254
  • € 458.00 / 100µg
Description
C1orf123 Polyclonal Antibody for Western Blot, IHC (P), ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1orf123
Polyclonal
Unconjugated
C1orf123
C1orf123; chromosome 1 open reading frame 123; CXXC motif containing zinc binding protein; CZIB; UPF0587 protein C1orf123
Rabbit
Protein G
RUO
54987
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
4 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q9NWV4
C1orf123
Recombinant Human UPF0587 protein C1orf123 protein (1-160AA).