missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf151 Monoclonal Antibody (OTI4H7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526031
This item is not returnable.
View return policy
Description
C1orf151 Monoclonal Antibody for Western Blot, IHC (P)
The exact function of C1orf151 remains unknown.
Specifications
| C1orf151 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5TGZ0 | |
| MINOS1 | |
| Full length human recombinant protein of C1orf151 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H7 | |
| Unconjugated | |
| MINOS1 | |
| 2310028O11Rik; C1orf151; MIC10; MICOS complex subunit MIC10; MICOS10; MINOS1; MIO10; mitochondrial contact site and cristae organizing system subunit 10; mitochondrial inner membrane organizing system 1; mitochondrial inner membrane organizing system protein 1; novel DUF543 domain containing protein; RGD1560187; RP5-1056L3.2; UPF0327 protein C1orf151; UPF0327 protein C1orf151 homolog | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 440574 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction