missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf83 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20940-1-AP-150UL
This item is not returnable.
View return policy
Description
TCEANC2 (Transcription Elongation Factor A N-Terminal And Central Domain Containing 2) is a Protein Coding gene.Specifications
| C1orf83 | |
| Polyclonal | |
| Unconjugated | |
| TCEANC2 | |
| C1orf83, RP4 758J24.3, TCEANC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127428, 366431, 66526 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIC7, Q8R2M0, Q96MN5 | |
| Tceanc2 | |
| C1orf83 Fusion Protein Ag15040 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction