missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C1orf86 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15756431 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15756431 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15756431 Supplier Invitrogen™ Supplier No. PA558555

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GGHLESPARS LPQRPAPDPC RAPRVEQQPS VEGAAALRSC PMCQKEFAPR LTQLDVDSHL AQCLAESTED VTW Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 55%.

C1orf86 is a component of the Fanconi anemia (FA) complex required to recruit the FA complex to DNA interstrand cross-links (ICLs) and promote ICLs repair. Following DNA damage recognizes and binds 'Lys-63'-linked ubiquitin generated by RNF8 at ICLs and recruits other components of the FA complex. It promotes translesion synthesis via interaction with REV1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1orf86
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAAP20
Gene Accession No. Q6NZ36
Gene Alias 2610002J02Rik; C1orf86; Faap20; FANCA-associated protein of 20 kDa; Fanconi anemia core complex associated protein 20; Fanconi anemia core complex-associated protein 20; Fanconi anemia-associated protein of 20 kDa; FP7162; likely orthologue of H. sapiens chromosome 1 open reading frame 86 (C1orf86); LOC362678; RGD1308923; RIKEN cDNA 2610002J02 gene
Gene Symbols FAAP20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C1orf86. Recombinant protein control fragment (Product #RP-94374).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.