missing translation for 'onlineSavingsMsg'
Learn More

C1QL1 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen C1QL1
Concentration 400 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30235217
View Documents
Proteintech
15757-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234761
View Documents
Proteintech
15757-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The C1QL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C1QL1. This antibody recognizes human, mouse, rat antigen. The C1QL1 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.
Specifications

Specifications

C1QL1
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
C1QRF; CRF
IgG
Antigen affinity purification
Polyclonal Antibody
400 μg/mL
Polyclonal
Liquid
RUO
10882
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.