Learn More
C1qR1/CD93 Antibody (273107), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB23791K
Product Code. 18513589
  • ONLINE EXCLUSIVE PRICE
    € 652.00
    € 616.35 / 0.10mL
    Save € 35.65
    5% Off
Description
C1qR1/CD93 Monoclonal antibody specifically detects C1qR1/CD93 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1qR1/CD93
Monoclonal
DyLight 488
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, CyTOF
273107
Western Blot, Flow Cytometry, CyTOF-ready
C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Mouse myeloma cell line NS0-derived recombinant human C1q R1/CD93, Ala24-Lys580, Accession # Q9NPY3
0.1 mL
B Cell Development and Differentiation Markers
22918
Store at 4°C in the dark.