missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qTNF5/CTRP5 Antibody (332923), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3167G
This item is not returnable.
View return policy
Description
C1qTNF5/CTRP5 Monoclonal antibody specifically detects C1qTNF5/CTRP5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Specifications
| C1qTNF5/CTRP5 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 332923 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP5/C1qTNF5, Ser16-Ala243 (Gln44Arg), Accession # Q9BXJ0 | |
| 0.1 mL | |
| Vision | |
| 114902 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction