Learn More
C1R Mouse anti-Canine, Human, Clone: OTI1F1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800085
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
C1r B chain is a serine protease that combines with C1q and C1s to form C1, the first component of the classical pathway of the complement system.Specifications
| C1R | |
| Monoclonal | |
| Unconjugated | |
| C1R | |
| EDSPD1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 477707, 715 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1F1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P00736 | |
| C1R | |
| Human recombit protein fragment corresponding to amino acids 262-562 of human C1R produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.