missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1S Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528253
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HepG2 culture medium pellet, HepG2 culture medium supernatant. Predicted reactivity: Rat (81%), Pig (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a serine protease, which is a major constituent of the human complement subcomponent C1. C1s associates with two other complement components C1r and C1q in order to yield the first component of the serum complement system. Defects in this gene are the cause of selective C1s deficiency.
Specifications
| C1S | |
| Polyclonal | |
| Unconjugated | |
| C1S | |
| AA959438; AI255193; AI327365; basic proline-rich peptide IB-1; C1 esterase; C1s; C1s1; C1sa; complement C1s; complement C1s subcomponent; Complement C1s subcomponent heavy chain; Complement C1s subcomponent light chain; Complement C1s-A subcomponent; Complement C1s-A subcomponent heavy chain; Complement C1s-A subcomponent light chain; Complement component 1 subcomponent s; complement component 1 subcomponent s-A; complement component 1, s subcomponent; complement component 1, s subcomponent 1; complement component C1SA; r-gsp; serine protease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.72 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P09871 | |
| C1S | |
| Recombinant protein encompassing a sequence within the center region of human C1s. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction