missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2, Mouse anti-Human, Clone: 2C11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000717-M15.100ug
This item is not returnable.
View return policy
Description
Sequence: APSCPQNVNISGGTFTLSHGWAPGSLLTYSCPQGLYPSPASRLCKSSGQWQTPGATRSLSKAVCKPVRCPAPVSFENGIYTPRLGSYPVGGNVSFECEDGFILRGSPVRQCRPNGMWDGETAVCDNGAGHCPNPGISLGAVRTGFRFGHGDKVRYRCSSNLVLTGSSERECQGNGVWSGTEPICRQPYSYDFPEDVAPALGTSFSHMLGATNPTQKTKESLGRSpecifications
| C2 | |
| Monoclonal | |
| Unconjugated | |
| complement component 2 | |
| CO2/DKFZp779M0311 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2C11 | |
| In 1x PBS, pH 7.4 | |
| NM_000063.3 | |
| C2 | |
| C2 (NP_000054.2, 21 a.a. ∼ 243 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 717 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction