missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119046
This item is not returnable.
View return policy
Description
C2 Polyclonal Antibody for Western Blot, IHC (P)
Component C2 is a serum glycoprotein that functions aspart of the classical pathway of the complement system. Activated C1 cleaves C2 into C2a and C2b. The serine proteinase C2a then combines with complement factor 4b to create the C3 or C5 convertase. Deficiency of C2 has been reported to associated with certain autoimmune diseases and SNPs in this gene have been associated with altered susceptibility to age-related macular degeneration. This gene localizes within the class III region of the MHC on the short arm of chromosome 6. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described in publications but their full-length sequence has not been determined.
Specifications
| C2 | |
| Polyclonal | |
| Unconjugated | |
| C2 | |
| ARMD14; C2; C3/C5 convertase; classical-complement pathway C3/C5 convertase; CO2; Complement C2; Complement C2a fragment; Complement C2b fragment; complement component 2; complement component 2 (within H-2S); complement component C2; complement factor C2; DADB-122G4.1; I79_016440; MHC complement component C2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 717 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P06681 | |
| C2 | |
| Recombinant protein Complement Component 2. The antigen corresponds to amino acid range 245-451 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction