missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C20orf10 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Product Code. 17909542
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909542 200 μL 200µL
17939882 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17909542 Supplier Invitrogen™ Supplier No. PA5145393

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

For immunocytochemistry the cells were formalin fixed.

May play a significant role in p53/TP53-mediating signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C20orf10
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene TP53TG5
Gene Accession No. Q9Y2B4
Gene Alias C20orf10; CLG01; P3H3; RGD1561934; TP53 target 5; TP53-inducible gene 5 protein; TP53-target gene 5 protein; TP53TG5
Gene Symbols TP53TG5
Host Species Rabbit
Immunogen Recombinant Human TP53TG5 protein, Tyr190-Thr262 (Accession #Q9Y2B4).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27296
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.