Learn More
DDX56 Monoclonal Antibody (OTI1G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802941
Encompass_Preferred
 View more versions of this product
Product Code. 17571783
  • € 540.87 / 100µL
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit.
Specifications
Specifications
DDX56
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NY93
DDX56
Human recombit protein fragment corresponding to amino acids 323-547 of human DDX56 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1G6
Unconjugated
DDX56
DDX21, DDX26, NOH61
Mouse
Affinity Chromatography
RUO
54606
-20° C, Avoid Freeze/Thaw Cycles