missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf3 Monoclonal Antibody (OTI1D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504500S
This item is not returnable.
View return policy
Description
C20orf3 Monoclonal antibody specifically detects C20orf3 in Human samples. It is validated for Western BlotSpecifications
| C20orf3 | |
| Monoclonal | |
| 0.57 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06203 | |
| Human recombinant protein fragment corresponding to amino acids 42-278 of human PPAT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| OTI1D5 | |
| Unconjugated | |
| Ppat | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5471 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction