missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf79 Monoclonal Antibody (OTI4D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812873
This item is not returnable.
View return policy
Description
SCP2D1 (SCP2 Sterol Binding Domain Containing 1) is a Protein Coding gene. Diseases associated with SCP2D1 include Corneal Dystrophy, Posterior Polymorphous, 1 and D-Bifunctional Protein Deficiency. An important paralog of this gene is HSD17B4.Specifications
| C20orf79 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJQ7 | |
| SCP2D1 | |
| Full length human recombit protein of human C20orf79 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4D10 | |
| Unconjugated | |
| SCP2D1 | |
| C20orf79, HSD22, dJ1068E13.2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 140856 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction