missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf79 Monoclonal Antibody (OTI4D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812873S
This item is not returnable.
View return policy
Description
C20orf79 Monoclonal antibody specifically detects C20orf79 in Human samples. It is validated for Western BlotSpecifications
| C20orf79 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UJQ7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 140856 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4D10 | |
| Unconjugated | |
| SCP2D1 | |
| 1700010M22Rik; C20orf79; dJ1068E13.2; HSD22; likely ortholog of H. sapiens chromosome 20 open reading frame 79 (C20orf79); RGD1308385; SCP2 sterol binding domain containing 1; SCP2 sterol-binding domain containing 1; SCP2 sterol-binding domain-containing protein 1; Scp2d1; sterol carrier protein 2-like protein | |
| Full length human recombinant protein of human C20orf79 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction