missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf59 Antibody (OTI2H5), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72060V
This item is not returnable.
View return policy
Description
C21orf59 Monoclonal specifically detects C21orf59 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| C21orf59 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFAP298 | |
| Full length recombinant protein of human C21orf59 (NP_067077) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2H5 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| C21orf48, chromosome 21 open reading frame 48, chromosome 21 open reading frame 59, FLJ20467, FLJ37137, FLJ40247, hypothetical protein LOC56683, kur, kurly | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 56683.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction