missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C21orf59 Monoclonal Antibody (OTI2E5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525158
This item is not returnable.
View return policy
Description
C21orf59 Monoclonal Antibody for Western Blot, ICC/IF, Flow
The function of the C21orf59 protein remains unknown.
Specifications
| C21orf59 | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P57076 | |
| C21ORF59 | |
| Full length human recombinant protein of C21orf59 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI2E5 | |
| Unconjugated | |
| C21ORF59 | |
| C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56683 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction