Learn More
C21orf59 Monoclonal Antibody (OTI2E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501348S
Encompass_Preferred
 View more versions of this product
Product Code. 17553981
  • € 242.00 / 30µL
Description
C21orf59 Monoclonal antibody specifically detects C21orf59 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
C21orf59
Monoclonal
0.75 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5I0X7
Mouse
Affinity chromatography
RUO
130502
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Flow Cytometry, Immunocytochemistry, Western Blot
OTI2E5
Unconjugated
Ttc32
C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59
Full length human recombinant protein of human TTC32 produced in HEK293T cell.