Learn More
C22orf25 Mouse, anti-Human, Clone: 2G5B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68625-1-IG-150UL
This item is not returnable.
View return policy
Description
The C22orf25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C22orf25. This antibody recognizes human antigen. The C22orf25 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| C22orf25 | |
| 2G5B8 | |
| C22orf25, DKFZp761P1121 | |
| 150 μL | |
| 128989 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |