Learn More
C22orf25 Mouse, anti-Human, Clone: 2G5B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68625-1-IG-20UL
 View more versions of this product
Product Code. 18097385
  • € 145.00 / 20µL
Description
The C22orf25 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C22orf25. This antibody recognizes human antigen. The C22orf25 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
C22orf25
2G5B8
C22orf25, DKFZp761P1121
20 μL
128989
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa