Learn More
Beskrivelse
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human A431 whole cell, human HL-60 whole cell, human U2OS whole cell, human PC-3 whole cell, rat lung tissue, mouse HEPA1-6 whole cell. Flow: HL-60 cell, PC-3 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Tekniske data
Tekniske data
| Antigen | C22orf29 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | RTL10 |
| Gene Accession No. | Q7L3V2 |
| Gene Alias | BH3-only protein; BOP; C22orf29; chromosome 22 open reading frame 29; protein Bop; Retrotransposon Gag-like protein 10; RTL10 |
| Gene Symbols | RTL10 |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
