missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2CD3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81963V
This item is not returnable.
View return policy
Description
C2CD3 Polyclonal antibody specifically detects C2CD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| C2CD3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 405 | |
| C2 calcium-dependent domain containing 3 | |
| C2CD3 antibody was raised against an 18 amino acid peptide near the center of human C2CD3. The immunogen is located within amino acids 1490 - 1540 of C2CD3. Amino Acid Squence: YGNDSVERPHQTDSWIGS | |
| 0.1 mL | |
| Signal Transduction | |
| 26005 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction