missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2CD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA572860
This item is not returnable.
View return policy
Description
Specificity: C2CD3 antibody is human, mouse and rat reactive.
C2CD3 Polyclonal antibody specifically detects C2CD3 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
Specifications
| C2CD3 | |
| Polyclonal | |
| Unconjugated | |
| C2CD3 | |
| AU020772; C2 calcium dependent domain containing 3; C2 calcium-dependent domain containing 3; C2 domain-containing protein 3; C2CD3; Hty; OFD14; Protein hearty; RGD1307366 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26005, 277939, 293148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q4AC94, Q52KB6 | |
| C2CD3 | |
| C2CD3 antibody for an 18 amino acid peptide near the center of human C2CD3. The immunogen is within amino acids 1490 - 1540 of C2CD3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction