missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2CD5 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74595B
This item is not returnable.
View return policy
Description
C2CD5 Polyclonal antibody specifically detects C2CD5 in Human samples. It is validated for Western Blot
Specifications
| C2CD5 | |
| Polyclonal | |
| Western Blot | |
| DKFZp779N2044, hypothetical protein LOC9847, KIAA0528 | |
| The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human KIAA0528 using the numbering given in entry NP_055617.1 (GeneID 9847). | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 9847 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction