missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf42 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48620
This item is not returnable.
View return policy
Description
C2orf42 Polyclonal antibody specifically detects C2orf42 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| C2orf42 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Simple Western reported by internal validation, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| chromosome 2 open reading frame 42 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HKSNASKDETAQRCIHFFACICAFASDETLAQEFSDFLNFDSSGLKEIIVPQLGCHSESTVSACESTASKSKKRRKDEVSG | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54980 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction