missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf62 Monoclonal Antibody (OTI2C6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505505S
This item is not returnable.
View return policy
Description
C2orf62 Monoclonal antibody specifically detects C2orf62 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| C2orf62 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z7H3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 375307 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2C6 | |
| Unconjugated | |
| CATIP | |
| C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437 | |
| Full length human recombinant protein of human C2orf62 produced in HEK239T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction