missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf62 Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505508
This item is not returnable.
View return policy
Description
Plays a role in primary ciliogenesis by modulating actin polymerization.Specifications
| C2orf62 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z7H3 | |
| CATIP | |
| Full length human recombit protein of human C2orf62 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D9 | |
| Unconjugated | |
| CATIP | |
| C2orf62 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 375307 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction