missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C2orf7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15760102 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15760102 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760102 Supplier Invitrogen™ Supplier No. PA562308

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DYLYFQVLSP GDIRYIFTAT PAKDFGGIFH TRYEQIHLVP AEPPEACGEL SNGFFIQDQI ALVERGGCSF LSKTRVVQ Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen C2orf7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRADC1
Gene Accession No. Q9BSG0
Gene Alias C2orf7; hPAP21; PAP21; PRADC1; protease associated domain containing 1; protease-associated domain containing 1; protease-associated domain-containing glycoprotein 21 kDa; protease-associated domain-containing protein 1; Protease-associated domain-containing protein of 21 kDa; UNQ833/PRO1760
Gene Symbols PRADC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C2orf7. Recombinant protein control fragment (Product #RP-101285).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.