missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: HSDLVNSVAK SGIQFLKKPK AIRPGCQGGS ERGSIPLLVK NSTCYDAGEG LAEEQPSPRR NQTTAKGLCQ LMGDPASGKR KDMEGLIPGT KTSSS Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 42%.
The protein encoded by this gene is highly expressed in photoreceptors and may associate with the primary cilium of the outer segment. The encoded protein appears to undergo post-translational lipid modification. Nonsense and missense variants of this gene appear to cause a recessive form of retinitis pigmentosa.
Specifications
Specifications
| Antigen | C2orf71 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | C2orf71 |
| Gene Accession No. | A6NGG8 |
| Gene Alias | C2orf71; chromosome 2 open reading frame 71; PCARE; Photoreceptor cilium actin regulator; RP54; uncharacterized protein C2orf71 |
| Gene Symbols | C2orf71 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
