Learn More
Tocris Bioscience™ C34
Description
C34 is a TLR4 inhibitor; attenuates TLR4 signaling in enterocytes and macrophages in vitro, and reduces systemic inflammation in mouse models of endotoxemia and necrotizing enterocolitis (NEC). Also suppresses LPS signaling in human ileum displaying NEC ex-vivo. Displays no significant effects on signaling via TLR2 or TLR9.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | C34 |
| Molecular Weight (g/mol) | 389.4 |
| Inhibitors | C34 |