missing translation for 'onlineSavingsMsg'
Learn More

C3a Receptor Mouse anti-Human, Brilliant Violet 650, Clone: hC3aRZ8, BD Optibuild™

Product Code. 15858299
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15858299 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15858299 Supplier BD Biosciences Supplier No. 743495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The hC3aRZ8 monoclonal antibody specifically binds to the human C3a Receptor (C3aR). C3aR is a seven-transmembrane glycoprotein, G-protein-coupled receptor that is the specific receptor for C3a anaphylatoxin. The C3aR consists of 482 amino acids forming a single polypeptide chain that is encoded by the C3AR1 /gene located on chromosome 12 (location 12p13.31). C3aR are expressed by eosinophils, basophils, neutrophils, dendritic cells, monocytes, macrophages, endothelial cells and some T cells. C3a is a bioactive cleavage product released from Complement Component 3 (C3) during complement activation. C3a plays a role in a variety of cellular immune responses as well as being a potent pro-inflammatory agent. In response to bound C3a, this receptor stimulates cellular responses including chemotaxis, granule enzyme release and superoxide anion production and causes increased vascular permeability. In vivo, C3a production can initiate, contribute to, or exacerbate the inflammatory reactions seen in gram-negative bacterial sepsis, trauma, ARDS, ischemic heart disease, post-dialysis syndrome, and several autoimmune diseases including rheumatoid arthritis, lupus erythematosus, and acute glomerulonephritis.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C3a Receptor
Classification Monoclonal
Clone hC3aRZ8
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias C3AR, C3a-R, C3AR1, C3a anaphylatoxin chemotactic receptor, AZ3B
Host Species Mouse
Immunogen Human C3aR
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.