missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C3orf10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543671
This item is not returnable.
View return policy
Description
Peptide sequence: YIEIITSSIK KIADFLNSFD MSCRSRLATL NEKLTALERR IEYIEARVTK Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 100%.
C3orf10 is involved in regulation of actin and microtubule organization. It is a part of a WAVE complex that activates the Arp2/3 complex.
Specifications
| C3orf10 | |
| Polyclonal | |
| Unconjugated | |
| BRK1 | |
| 6720456B07Rik; AW011779; BRICK1 subunit of SCAR/WAVE actin nucleating complex; BRICK1, SCAR/WAVE actin nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit, homolog; BRK1; C22H3orf10; C3orf10; haematopoietic stem/progenitor cell protein 300; hHBrk1; HSPC300; MDS027; Probable protein BRICK1; protein BRICK1; protein BRICK1-like protein; zgc:86903 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55845 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WUW1 | |
| BRK1 | |
| Synthetic peptide directed towards the middle region of human C3orf10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction