missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf15 Monoclonal Antibody (OTI6G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802361S
This item is not returnable.
View return policy
Description
C3orf15 Monoclonal antibody specifically detects C3orf15 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| C3orf15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N556 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 60312 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6G1 | |
| Unconjugated | |
| Afap1 | |
| 4932425I24Rik; AAT1; AAT-1; AAT1alpha; AAT1-alpha; AMY-1-associating protein expressed in testis 1; AMY-1-associating protein expressed in testis 1 homolog; C3orf15; CaM-IP2; CFAP91; cilia and flagella associated protein 91; cilia- and flagella-associated protein 91; Maats1; MYCBP associated and testis expressed 1; MYCBP/AMY-1-associated testis-expressed protein 1; MYCBP/AMY-1-associated, testis expressed 1; MYCBP-associated, testis expressed 1; MYCBP-binding protein; protein MAATS1; RGD1305539; SPATA26; spermatogenesis associated 26 | |
| Human recombinant protein fragment corresponding to amino acids 100-386 of human AFAP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction