missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf15 Mouse anti-Human, Clone: OTI4F2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802372
This item is not returnable.
View return policy
Description
C3orf15 (MAATS1, MYCBP Associated And Testis Expressed 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include adenosylmethionine decarboxylase activity.Specifications
| C3orf15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z4T9 | |
| MAATS1 | |
| Human recombit protein fragment corresponding to amino acids 1-313 of human C3orf15 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4F2 | |
| Unconjugated | |
| MAATS1 | |
| AAT1, AAT1alpha, C3orf15, CFAP91, CaM-IP2, SPATA26 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89876 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction