missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf20 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20906-1-AP-150UL
This item is not returnable.
View return policy
Description
20906-1-AP can recognize both 88 and 101 kDa isoforms.
The function of C3orf20 has not been widely studied, and is yet to be fully elucidated. Two isoforms were produced by alternative splicing with predicted MW of 88 and 101 kDa according to UniProt.Specifications
| C3orf20 | |
| Polyclonal | |
| Unconjugated | |
| C3orf20 | |
| C3orf20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84077 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8ND61 | |
| C3orf20 | |
| C3orf20 Fusion Protein Ag15031 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction