missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf36 Antibody (OTI8G4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72218C
This item is not returnable.
View return policy
Description
C3orf36 Monoclonal specifically detects C3orf36 in Human samples. It is validated for Western Blot.
Specifications
| C3orf36 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| C3orf36 | |
| Full length recombinant protein of human C3orf36 (NP_079317) produced in E.coli. | |
| 0.1 mL | |
| 80111.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI8G4 | |
| Western Blot | |
| C3orf36 chromosome 3 open reading frame 36 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction