missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf38 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00285237-B01P
This item is not returnable.
View return policy
Description
C3orf38 Polyclonal antibody specifically detects C3orf38 in Human samples. It is validated for Western Blot
Specifications
| C3orf38 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| chromosome 3 open reading frame 38, FLJ54270, hypothetical protein LOC285237, MGC26717 | |
| C3orf38 (NP_776185.1, 1 a.a. - 327 a.a.) full-length human protein. MSGLSFSEMEGCRNLLGLLDNDEIMALCDTVTNRLVQPQDRQDAVHAILAYSQSAEELLRRRKVHREVIFKYLATQGIVIPPATEKHNLIQHAKDYWQKQPQLKLKETPEPVTKTEDIHLFQQQVKEDKKAEKVDFRRLGEEFCHWFFGLLNSQNPFLGPPQDEWGPQHFWHDVKLRFYYNTSEQNVMDYHGAEIVSLRLLSLVKEEFLFLSPNLDSHGLKCASSPHGLVMVGVAGTVHRGNTCLGIFEQIFGLIRCPFVENTWKIKFINLKIMGESSLAPGTLPKPSVKFEQSDLEAFYNVITVCGTNEVRHNVKQASDSGTGDQV | |
| 0.05 mg | |
| Lipid and Metabolism | |
| 285237 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_776185.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction