missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C4orf22 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560347
This item is not returnable.
View return policy
Description
Immunogen sequence: QEEGLKALDN IVTQFNAYED FLDSQITTVD LYYLEDETLA RQLVELGYRG TGERVKREDF EARKAAIEIA RLAERAQQKT LTSAGK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 31%.
C4orf22 is a protein coding gene.
Specifications
| C4orf22 | |
| Polyclonal | |
| Unconjugated | |
| C4orf22 | |
| C4orf22; CFAP299; chromosome 4 open reading frame 22; Cilia- and flagella-associated protein 299; uncharacterized protein C4orf22 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 255119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6V702 | |
| C4orf22 | |
| Recombinant protein corresponding to Human C4orf22. Recombinant protein control fragment (Product #RP-100295). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction