missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf36 Antibody (OTI7A7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72388AF594
This item is not returnable.
View return policy
Description
C4orf36 Monoclonal specifically detects C4orf36 in Human samples. It is validated for Western Blot.
Specifications
| C4orf36 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| C4orf36 | |
| Full length recombinant protein of human C4orf36 (NP_653246) produced in E.coli. | |
| 0.1 mL | |
| 132989.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI7A7 | |
| Western Blot | |
| chromosome 4 open reading frame 36, hypothetical protein LOC132989, MGC26744 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction