missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf36 Monoclonal Antibody (OTI2H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808367
This item is not returnable.
View return policy
Description
C4orf36 (Chromosome 4 Open Reading Frame 36) is a Protein Coding gene. An important paralog of this gene is ENSG00000285458.Specifications
| C4orf36 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96KX1 | |
| C4orf36 | |
| Full length human recombit protein of human C4orf36 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2H5 | |
| Unconjugated | |
| C4orf36 | |
| chromosome 4 open reading frame 36 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 132989 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction